1,1,3,3-Tetramethyldisiloxane

235733-100G 235733-25G
220046-477EA 80.31 USD
220046-477 220046-478
1,1,3,3-Tetramethyldisiloxane
1,1,3,3-Tetramethyldisiloxane
Formula: C₄H₁₄OSi₂
MW: 134.33 g/mol
Boiling Pt: 71 °C (1013 hPa)
Density: 0.7555 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008256
CAS Number: 3277-26-7
EINECS: 221-906-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR