Di-tert-butylcarbodiimide

Synonyms: 1,3-Di-tert-butylcarbodiimide, N,N'-Di-tert-butylcarbodiimide, N,N-di-tert-butylcarbodiimide, N,N'-methanetetraylbis(1,1-dimethylethylamine)

235563-5G 235563-1G
220046-456EA 294 USD
220046-456 220046-455
Di-tert-butylcarbodiimide
Di-tert-butylcarbodiimide
Formula: C₉H₁₈N₂
MW: 154.26 g/mol
MDL Number: MFCD00010231
CAS Number: 691-24-7
EINECS: 211-719-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR