2-Benzylaniline ≥99%

Synonyms: 2-Aminodiphenylmethane, 2-Benzylaniline

235350-10G 235350-50G
220046-218EA 173.38 USD
220046-218 220046-219
2-Benzylaniline ≥99%
2-Benzylaniline
Formula: C₁₃H₁₃N
MW: 183.25 g/mol
MDL Number: MFCD00007750
CAS Number: 28059-64-5
EINECS: 248-806-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR