(α,α,α-Trifluoro-2-tolyl)acetic acid ≥98%

Warning

Synonyms: 2-(Trifluoromethyl)phenylacetic acid, (2-(Trifluoromethyl)phenyl)acetic acid

235296-1G 235296-5G
220046-214EA 29.4 USD
220046-214 220046-215
(α,α,α-Trifluoro-2-tolyl)acetic acid ≥98%
(α,α,α-Trifluoro-2-tolyl)acetic acid
Formula: F₃CC₆H₄CH₂COOH
MW: 204.15 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 98…100 °C
MDL Number: MFCD00004327
CAS Number: 3038-48-0
EINECS: 221-240-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR