Methyl (trimethylsilyl)acetate ≥97%

Synonyms: (Trimethylsilyl)acetic acid methyl ester

234966-5G 234966-25G
220045-735EA 66.92 USD
220045-735 220045-734
Methyl (trimethylsilyl)acetate ≥97%
Methyl (trimethylsilyl)acetate
Formula: C₆H₁₄O₂Si
MW: 146.26 g/mol
CAS Number: 2916-76-9
EINECS: 220-845-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR