Butyl acrylate ≥99%

Synonyms: n-Butyl acrylate, Acrylic acid butyl ester

234923-100ML 234923-1L 234923-18L
220045-726EA 71.66 USD
220045-726 220045-728 220045-727
Butyl acrylate ≥99%
Butyl acrylate
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
MDL Number: MFCD00008809
CAS Number: 141-32-2
EINECS: 205-480-7
UN: 2348
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR