Tributyltin hydride

Danger

Synonyms: Tri-n-butyltin hydride

234788-10G 234788-500G 234788-50G
220046-166EA 42.85 USD
220046-166 220046-167 220046-168
Tributyltin hydride
Tributyltin hydride
Formula: C₁₂H₂₈Sn
MW: 291.06 g/mol
Boiling Pt: 112…114 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00009416
CAS Number: 688-73-3
EINECS: 211-704-4
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR