Methyl-3-chlorobenzoate ≥98%

Synonyms: 3-Chlorobenzoic acid emthyl ester, Methyl 3-chlorobenzoate

234621-25G 234621-5G
220046-156EA 109.85 USD
220046-156 220046-157
Methyl-3-chlorobenzoate ≥98%
Methyl-3-chlorobenzoate
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: 21 °C
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00000592
CAS Number: 2905-65-9
EINECS: 220-814-1
Merck Index: 13,02142

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR