Methyl laurate ≥99.5%

Synonyms: Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester

234591-5G 234591-2.5G 234591-25G
220046-155EA 45.66 USD
220046-155 220046-153 220046-154
Methyl laurate ≥99.5%
Methyl laurate
Formula: C₁₃H₂₆O₂
MW: 214.35 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 4.5 °C
Density: 0.8694 g/cm³ (20 °C)
MDL Number: MFCD00008966
CAS Number: 111-82-0
EINECS: 203-911-3

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR