5-tert-Butyl-m-xylene ≥98%

Synonyms: 1-tert-Butyl-3,5-dimethylbenzene, 1-(tert-Butyl)-3,5-dimethylbenzene

233587-100G 233587-500G
220046-322EA 56 USD
220046-322 220046-323
5-tert-Butyl-m-xylene ≥98%
5-tert-Butyl-m-xylene
Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –22 °C
Density: 0.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008832
CAS Number: 98-19-1
EINECS: 202-647-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR