Acetic acid-[D₄]

Danger

Synonyms: Ethanoic acid-D4, Acetic-D3 acid-D, Acetic-d3 acid-d, Acetic acid-[²H₄], Tetradeuteroacetic acid, Acetic acid-d₄

233315-25G 233315-5G
220046-294EA 49.23 USD
220046-294 220046-295
Acetic acid-[D₄]
Acetic acid-[D4]
Formula: D₃CCOOD
MW: 64.02 g/mol
Boiling Pt: 115.5 °C (1013 hPa)
Melting Pt: 16 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00051051
CAS Number: 1186-52-3
EINECS: 214-693-4
UN: 2789
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR