2,6-Difluoropyridine

Danger

233277-5G 233277-25G
220046-290EA 48.57 USD
220046-290 220046-289
2,6-Difluoropyridine
2,6-Difluoropyridine
Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 123…124 °C (1013 hPa)
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006227
CAS Number: 1513-65-1
EINECS: 216-152-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR