2,4-Difluorophenol ≥99%

233269-25G 233269-5G
220046-286EA 326.31 USD
220046-286 220046-287
2,4-Difluorophenol ≥99%
2,4-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: 20 °C
Density: 1.36 g/cm³ (20 °C)
MDL Number: MFCD00009715
CAS Number: 367-27-1
EINECS: 206-688-0
UN: 2921
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR