1,3-Difluoro-4-nitrobenzene ≥99%

Synonyms: 2,4-Difluoro-1-nitrobenzene, 2,4-Difluoronitrobenzene

233226-100G 233226-25G
220046-280EA 127.08 USD
220046-280 220046-281
1,3-Difluoro-4-nitrobenzene ≥99%
1,3-Difluoro-4-nitrobenzene
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 10 °C
Density: 1.4571 g/cm³ (14 °C)
MDL Number: MFCD00007050
CAS Number: 446-35-5
EINECS: 207-167-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR