2,6-Bis(trifluoromethyl)benzoic acid ≥98%

233196-5G 233196-1G
220046-278EA 168 USD
220046-278 220046-277
2,6-Bis(trifluoromethyl)benzoic acid ≥98%
2,6-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 138 - 139 °C
MDL Number: MFCD00000376
CAS Number: 24821-22-5
EINECS: 246-479-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR