4'-(Trifluoromethyl)acetophenone ≥98%

Warning

Synonyms: 1-[4-(Trifluoromethyl)phenyl]ethan-1-one, 4-(Trifluoromethyl)acetophenone, p-(Trifluoromethyl)acetophenone

233188-25G 233188-5G
220046-274EA 85.83 USD
220046-274 220046-275
4'-(Trifluoromethyl)acetophenone ≥98%
4'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 26…29 °C
Density: 0.924 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7
EINECS: 211-913-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR