3'-(Trifluoromethyl)acetophenone ≥99%

Warning

Synonyms: m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one

233161-25G 233161-5G
220046-138EA 55.92 USD
220046-138 220046-271
3'-(Trifluoromethyl)acetophenone ≥99%
3'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 198 °C (1013 hPa)
Density: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000391
CAS Number: 349-76-8
EINECS: 206-490-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR