2-Chloroaniline

Danger

23300-500ML 23300-100ML
220045-050EA 120 USD
220045-050 220045-049
2-Chloroaniline
2-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.213 g/cm³ (20 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00007656
CAS Number: 95-51-2
EINECS: 202-426-4
UN: 2019
ADR: 6.1,II
Merck Index: 13,02136

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR