2',6'-Difluoroacetophenone ≥97%

Synonyms: 1-(2,6-Difluorophenyl)ethan-1-one, 1-(2,6-Difluorophenyl)ethanone

232890-1G 232890-5G
220046-129EA 29.72 USD
220046-129 220046-130
2',6'-Difluoroacetophenone ≥97%
2',6'-Difluoroacetophenone
Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 76…79 °C (15 mmHg)
Density: 1.19 g/cm³ (20 °C)
MDL Number: MFCD00000328
CAS Number: 13670-99-0
EINECS: 237-151-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR