3,5-Bis(trifluoromethyl)benzoic acid ≥98%

232882-5G 232882-25G
220046-128EA 37.69 USD
220046-128 220046-127
3,5-Bis(trifluoromethyl)benzoic acid ≥98%
3,5-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 140…144 °C
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00000388
CAS Number: 725-89-3
EINECS: 211-970-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR