3-Formylbenzoic acid ≥97%

Synonyms: Isophthalaldehydic acid, 3-Carboxybenzaldehyde

232793-5G 232793-1G
220046-121EA 152.92 USD
220046-121 220046-120
3-Formylbenzoic acid ≥97%
3-Formylbenzoic acid
Formula: C₈H₆O₃
MW: 150.13 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 175 °C
MDL Number: MFCD00039575
CAS Number: 619-21-6
EINECS: 210-585-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR