1-Bromo-2-chloroethane ≥98%

Synonyms: Ethylene chlorobromide

232750-5G 232750-250G
220046-114EA 33.92 USD
220046-114 220046-113
1-Bromo-2-chloroethane ≥98%
1-Bromo-2-chloroethane
Formula: BrH₂CCH₂Cl
MW: 143.41 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: –17 °C
Density: 1.7392 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000962
CAS Number: 107-04-0
EINECS: 203-456-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR