3-Hydroxy-2-nitro-6-picoline

Synonyms: 6-Methyl-2-nitropyridin-3-ol, 6-Methyl-2-nitro-3-pyridone, 3-Hydroxy-6-methyl-2-nitropyridine, 6-Metil-2-nitropyridine-3-ol

232742-10G 232742-50G
220046-111EA 154 USD
220046-111 220046-112
3-Hydroxy-2-nitro-6-picoline
3-Hydroxy-2-nitro-6-picoline
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 105…106 °C
MDL Number: MFCD00006259
CAS Number: 15128-90-2
EINECS: 239-192-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR