Ethyl methoxyacetate ≥97%

Synonyms: Methoxyacetic ethyl ester

232599-10G 232599-50G
220046-033EA 71.62 USD
220046-033 220046-034
Ethyl methoxyacetate ≥97%
Ethyl methoxyacetate
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 44 °C (1013 hPa)
Density: 1.0118 g/cm³ (15 °C)
MDL Number: MFCD00009176
CAS Number: 3938-96-3
EINECS: 223-521-7
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR