N-(Chloromethyl)phthalimide

Synonyms: 2-(Chloromethyl)isoindoline-1,3-dione

232424-10G 232424-50G
220046-025EA 108.77 USD
220046-025 220046-026
N-(Chloromethyl)phthalimide
N-(Chloromethyl)phthalimide
Formula: C₉H₆ClNO₂
MW: 195.6 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 123…126 °C
MDL Number: MFCD00005898
CAS Number: 17564-64-6
EINECS: 241-541-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR