Cesium hydroxide 50% (w/w) in aqueous solution

232068-100G 232068-500G
220045-990EA 186.15 USD
220045-990 220045-991
Cesium hydroxide 50% (w/w) in aqueous solution
Cesium hydroxide
Formula: CsOH
MW: 149.91 g/mol
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8,II
Merck Index: 13,02022

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR