Cesium hydroxide ≥99.9% in aqueous solution

232041-200G 232041-10G 232041-50G
220045-986EA 566.15 USD
220045-986 220045-985 220045-987
Cesium hydroxide ≥99.9% in aqueous solution
Cesium hydroxide
Formula: CsOH
MW: 149.91 g/mol
MDL Number: MFCD00010964
CAS Number: 21351-79-1
EINECS: 244-344-1
UN: 2681
ADR: 8,II
Merck Index: 13,02022

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR