Tetrachloro-p-benzoquinone

Warning

Synonyms: Tetrachloroquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 4-Chloranil, p-Chloranil, 2,3,5,6-Tetrachlorobenzo-1,4-quinone, Tetrachloro-1,4-benzoquinone

232017-25G 232017-100G
220045-980EA 92.06 USD
220045-980 220045-979
Tetrachloro-p-benzoquinone
Tetrachloro-p-benzoquinone
Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 290…293 °C
Density: 1.97 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001594
CAS Number: 118-75-2
EINECS: 204-274-4
Merck Index: 13,02088

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR