Antimony(III) oxide ≥99%, ReagentPlus®

Warning

Synonyms: Antimony trioxide, Diantimony trioxide, di-Antimony trioxide

230898-100G 230898-1KG
220045-951EA 59.54 USD
220045-951 220045-952
Antimony(III) oxide ≥99%, ReagentPlus®
Antimony (III) oxide
Formula: O₃Sb₂
MW: 291.52 g/mol
Boiling Pt: 1425 °C (1013 hPa)
Melting Pt: 654 °C
Density: 5.2 g/cm³ (20 °C)
MDL Number: MFCD00011214
CAS Number: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1,III
Merck Index: 13,00717

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR