4-Phenoxyphenol ≥99%

Synonyms: 4-Hydroxydiphenyl ether, Hydroquinone monophenyl ether

230669-25G 230669-5G
220045-927EA 218.62 USD
220045-927 220045-928
4-Phenoxyphenol ≥99%
4-Phenoxyphenol
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 80…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00002331
CAS Number: 831-82-3
EINECS: 212-611-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR