Methyl-4-cyanobenzoate ≥99%

Synonyms: 4-Cyanobenzoic acid methyl ester

230634-25G 230634-5G
220045-923EA 367.23 USD
220045-923 220045-924
Methyl-4-cyanobenzoate ≥99%
Methyl-4-cyanobenzoate
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 67 °C
MDL Number: MFCD00001823
CAS Number: 1129-35-7
EINECS: 214-443-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR