Sodium ethoxide 21% (w/w) in ethanol

Synonyms: Sodium ethylate, Sodium ethanolate

230553-100ML 230553-500ML
220045-919EA 29.46 USD
220045-919 220045-920
Sodium ethoxide 21% (w/w) in ethanol
Sodium ethoxide
Formula: C₂H₅NaO
MW: 68.05 g/mol
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3274
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR