Magnesium sulfate heptahydrate ≥98% ACS

230391-2.5KG 230391-25G 230391-500G 230391-5KG 230391-1KG
220045-899EA 185.23 USD
220045-899 220045-900 220045-901 220045-902 220045-898
Magnesium sulfate heptahydrate ≥98% ACS
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
Melting Pt: 1124 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR