Cobalt(II) nitrate hexahydrate, Reagent Grade

Danger

Synonyms: Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate

230375-500G 230375-100G
220045-897EA 158.77 USD
220045-897 220045-896
Cobalt(II) nitrate hexahydrate, Reagent Grade
Cobalt(II) nitrate hexahydrate
Formula: Co(NO₃)₂·6H₂O
MW: 291.03 g/mol
Melting Pt: 57 °C
Density: 1.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,III
Merck Index: 13,02469

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR