Methyl (S)-(-)-lactate ≥98%

Danger

Synonyms: (S)-Methyl 2-hydroxypropanoate, Methyl (S)-(-)-2-hydroxypropionate, Sodium carboxymethyl cellulose

230340-100G 230340-5G
220045-059EA 45.23 USD
220045-059 220045-060
Methyl (S)-(-)-lactate ≥98%
Methyl (S)-(-)-lactate
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: –44 °C
Density: 1.09 g/cm³ (20 °C)
MDL Number: MFCD00064265
CAS Number: 27871-49-4
EINECS: 248-704-9
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR