Tetrabutylammonium hydroxide 1 M in methanol

230189-100ML 230189-800ML 230189-1L 230189-250ML
220045-318EA 213.85 USD
220045-318 220045-321 220045-319 220045-320
Tetrabutylammonium hydroxide 1 M in methanol
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR