Tetrabutylammonium tetrahydroborate

Synonyms: Tetra-n-butylammonium borohydride, Tetrabutylammonium borohydride, Tetrabutylammonium tetrahydridoborate

230170-50G 230170-10G
220045-316EA 186.92 USD
220045-316 220045-315
Tetrabutylammonium tetrahydroborate
Tetrabutylammonium tetrahydroborate
Formula: C₁₆H₄₀BN
MW: 257.31 g/mol
Melting Pt: 124 - 126 °C
MDL Number: MFCD00012035
CAS Number: 33725-74-5
EINECS: 251-658-2
UN: 3131
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR