Oxazole

230138-1G 230138-10G
220045-312EA 124.92 USD
220045-312 220045-311
Oxazole
Oxazole
Formula: C₃H₃NO
MW: 69.06 g/mol
Boiling Pt: 68 - 69 °C (1013 hPa)
Melting Pt: -87 °C
Density: 1.08 g/cm³ (15 °C)
MDL Number: MFCD00009751
CAS Number: 288-42-6
EINECS: 206-020-8
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR