Antimony(III) potassium oxitartrate hydrate ≥99.95% (trace metals basis)

Warning

Synonyms: (R,R)-(+)-Antimony potassium tartrate hydrate, (+)-Antimony potassium L-tartrate hydrate, L(+)-Tartaric acid antimony potassium salt hydrate, L(+)-Antimony potassium tartrate hydrate, Antimony potassium L(+)-tartrate hydrate, (R,R)-(+)-Tartaric acid antimony potassium salt hydrate, Antimony potassium tartrate hydrate, Potassium antimony(III) tartrate hydrate

230057-100G 230057-10G
220045-304EA 1104.62 USD
220045-304 220045-305
Antimony(III) potassium oxitartrate hydrate ≥99.95% (trace metals basis)
Antimony(III) potassium oxitartrate hydrate
Formula: C₈H₄K₂O₁₂Sb₂·H₂O
MW: 631.84 g/mol (anhy)
MDL Number: MFCD00148863
CAS Number: 331753-56-1
EINECS: 234-293-3
UN: 1551
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR