di-Sodium tetraborate ≥99.998%

Danger

Synonyms: Borax, Sodium tetraborate

229946-25G 229946-100G 229946-5G
220045-093EA 60.57 USD
220045-093 220045-092 220045-094
di-Sodium tetraborate ≥99.998%
di-Sodium tetraborate
Formula: Na₂B₄O₇
MW: 201.22 g/mol
Boiling Pt: 1575 °C (1013 hPa)
Melting Pt: 742 °C
Density: 2.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081185
CAS Number: 1330-43-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 12,08733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR