Sodium bromide ≥99.99%

229881-10G 229881-100G
220045-084EA 119.38 USD
220045-084 220045-083
Sodium bromide ≥99.99%
Sodium bromide
Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1396 °C (1013 hPa)
Melting Pt: 753 °C
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6
EINECS: 231-599-9
Merck Index: 13,08666

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR