Selenium ≥99.99%

Danger

229865-5G 229865-100G 229865-20G
220045-080EA 95.23 USD
220045-080 220045-078 220045-079
Selenium ≥99.99%
Selenium
Formula: Se
Boiling Pt: 684.9 °C (1013 hPa)
Melting Pt: 217 °C
Density: 4.81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3283
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR