Potassium fluoride ≥99.99%

Danger

229814-10G 229814-100G
220045-881EA 204.62 USD
220045-881 220045-880
Potassium fluoride ≥99.99%
Potassium fluoride
Formula: KF
MW: 58.1 g/mol
Boiling Pt: 1505 °C (1013 hPa)
Melting Pt: 860 °C
Density: 2.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 13,07714

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR