Lithium nitrate ≥99.99%

Warning

229741-25G 229741-125G
220045-870EA 177.92 USD
220045-870 220045-869
Lithium nitrate ≥99.99%
Lithium nitrate
Formula: LiNO₃
MW: 68.95 g/mol
Melting Pt: 255 °C
Density: 2.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011094
CAS Number: 7790-69-4
EINECS: 232-218-9
UN: 2722
ADR: 5.1,III
Merck Index: 13,05558

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR