Lithium bromide ≥99.995%

Warning

229733-125G 229733-25G
220045-867EA 884.62 USD
220045-867 220045-868
Lithium bromide ≥99.995%
Lithium bromide
Formula: LiBr
MW: 86.84 g/mol
Boiling Pt: 1265 °C (1013 hPa)
Melting Pt: 550 °C
Density: 3.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011077
CAS Number: 7550-35-8
EINECS: 231-439-8
Merck Index: 13,05548

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR