Copper(II) acetate monohydrate ≥99.99%

Danger

Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

229601-50G 229601-10G
220045-848EA 1152.31 USD
220045-848 220045-847
Copper(II) acetate monohydrate ≥99.99%
Copper(II) acetate monohydrate
Formula: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Melting Pt: 115 °C
Density: 1.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149570
CAS Number: 6046-93-1
EINECS: 205-553-3
UN: 3077
ADR: 9,III
Merck Index: 13,02651

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR