Aluminium nitrate nonahydrate ≥99.997%

Warning

Synonyms: Aluminum nitrate nonahydrate

229415-100G 229415-10G
220045-827EA 670.77 USD
220045-827 220045-828
Aluminium nitrate nonahydrate ≥99.997%
Aluminium nitrate nonahydrate
Formula: Al(NO₃)₃·9H₂O
MW: 375.13 g/mol
Melting Pt: 73 °C
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS Number: 7784-27-2
EINECS: 236-751-8
UN: 1438
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.997%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR