Indigo

Warning

Synonyms: Pigment Blue 66, Vat Blue 1, Indigotin, 2,2'-Bis(2,3-dihydro-3-oxoindolyliden), Indigo (synthetic), 2-(1,3-Dihydro-3-oxo-2H-indol-2-ylidene)-1,2-dihydro-3H-indol-3-one

229296-100G 229296-25G
220045-818EA 211.4 USD
220045-818 220045-819
Indigo
Indigo
Formula: C₁₆H₁₀N₂O₂
MW: 262.27 g/mol
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005722
CAS Number: 482-89-3
EINECS: 207-586-9
Merck Index: 12,04977
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e4c344ac5f2c955a7b20d49372b316ace2bc416&vid=ab00958afc3ada9afe8bc18f4ce4da9f20e0b308&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR