Tributyltin methanolate

Danger

Synonyms: Tri-n-butyltin methoxide, Methoxytributylstannane, Tributyltin methoxide

229245-100G 229245-25G
220045-607EA 31.72 USD
220045-607 220045-608
Tributyltin methanolate
Tributyltin methanolate
Formula: C₁₃H₃₀OSn
MW: 321.09 g/mol
Boiling Pt: 97 °C (1013 hPa)
Density: 1.129 g/cm³ (20 °C)
MDL Number: MFCD00009419
CAS Number: 1067-52-3
EINECS: 213-933-5
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR