Diphenyltin dichloride

Synonyms: Dichlorodiphenylstannane

229202-5G 229202-25G
220045-603EA 21.88 USD
220045-603 220045-602
Diphenyltin dichloride
Diphenyltin dichloride
Formula: C₁₂H₁₀Cl₂Sn
MW: 343.83 g/mol
MDL Number: MFCD00000516
CAS Number: 1135-99-5
EINECS: 214-496-3
UN: 3146
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR